Learn More
Labconco™ General Glove Box: Oxygen and Moisture Removal Column
SDP
Catalog No. 16108380 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box, Controlled Atmosphere Glove Box
Catalog No. For Use With (Equipment)
16-108-380 Precise Basic Glove Box, Controlled Atmosphere Glove Box
1 options
Catalog No. 16-108-380 Supplier Labconco™ Supplier No. 5244200
Add to Cart
Add to Cart
Accessories for Labconco General Glove Box
  • For use with Labconco General Glove Box
  • Provides less than 5ppm oxygen and less than 50ppm moisture (refillable)
Specifications
For Use With (Equipment) Precise Basic Glove Box, Controlled Atmosphere Glove Box
Weight (English) 26 lb.
Weight (Metric) 11.8 Kg
Product Type Oxygen, Moisture Removal Column
WARNING: Cancer - www.P65Warnings.ca.gov