Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Glass Trap Insert for CentriVap™ System
GSA_VASDP

Catalog No. 16108366 Shop All Labconco Corporation Products
Change view
Click to view available options
Capacity (Metric):
2550 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
16-108-366 2550 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-108-366 Supplier Labconco™ Supplier No. 7397605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects CentriVap Cold Trap reservoir and lid from corrosives.

  • Included with CentriVap Acid Systems; accessory for other systems
  • Not compatible with pre-2005 Cold Traps

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Trap Insert
Capacity (Metric) 2550 mL
No. of Shelves 0
Material Glass
For Use With (Equipment) CentriVap™ concentrator cold trap
Weight (English) 4 lb.
Weight (Metric) 1.8 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.