Learn More
Labconco™ Hard Duct Exhaust Connector for Precise™ Glove Boxes
SDP
Catalog No. 16108413 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise HEPA Filtered Glove Box, Xpert Weigh Box
Catalog No. For Use With (Equipment)
16-108-413 Precise HEPA Filtered Glove Box, Xpert Weigh Box
1 options
Catalog No. 16-108-413 Supplier Labconco™ Supplier No. 5230400
Add to Cart
Add to Cart
Accessories for use with Precise Glove Boxes
Epoxy-coated steel, 6 in. diameter, for connecting the glove box to ductwork attached to a remote blower. Remote blower must be balanced with glove box's built-in blower. Requires ductwork and remote blower. For use with Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes.
Specifications
For Use With (Equipment) Precise HEPA Filtered Glove Box, Xpert Weigh Box
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Product Type Hard Duct Exhaust Connector
For Use With (Application) Connecting the glove box to a remote blower, when by-passing the glove box blower
WARNING: Cancer - www.P65Warnings.ca.gov