Learn More
Labconco™ Lids for CentriVap™Cold Traps
SDP
Catalog No. 10269685 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
CentriVap Concentrator Cold Trap
Catalog No. For Use With (Equipment)
10-269-685 CentriVap Concentrator Cold Trap
1 options
Catalog No. 10-269-685 Supplier Labconco™ Supplier No. 7386700
Add to Cart
Add to Cart
Provides corrosion resistance.
Stainless Steel Lid may be installed in place of the acrylic lid on -50 CentriVap Cold Trap when applications involve solvents which attack acrylic. Included on CentriVap Solvent and Proteomic Systems and -84 and -105 CentriVap Cold Traps.
Specifications
Material Stainless Steel
Type Concentrator Cold Trap Lid
For Use With (Equipment) CentriVap Concentrator Cold Trap