Learn More
Labconco™ Detergent Dispenser Kit
SDP
Catalog No. 10359205 Shop All Labconco Corporation Products
Change view
Click to view available options
Liquid Detergent Dispenser Kit
Catalog No.
10-359-205 Liquid Detergent Dispenser Kit
1 options
Catalog No. 10-359-205 Supplier Labconco™ Supplier No. 4587500
Add to Cart
Add to Cart
Includes
Peristaltic pump, float switch, tubing, one gallon (3.8L) plastic container and installation instructions
Automates liquid detergent dispensing
  • Connect to any SteamScrubber™ or FlaskScrubber™ glassware scrubber
Order Info
Detergent not included
Specifications
Product Type Liquid Detergent Dispenser Kit
For Use With (Application) To automate liquid detergent dispensing
For Use With (Equipment) FlaskScrubber™, SteamScrubber™ glassware washer
Includes Peristaltic pump, float switch, tubing, 1 gal. plastic container and installation instructions
Weight (English) Shipping 10 lb.
Weight (Metric) Shipping 4.5 kg
Quantity 1 Ea.
WARNING: Cancer - www.P65Warnings.ca.gov