Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Manual and Electric Hydraulic Lift Base Stands
Offers infinitely adjustable height from 25.5 and 33.5 in. (648 to 851mm)
Supplier: Labconco™ 3780100
Description
- Height adjustment on manual unit can be done via hand crank. Height adjustment on electric units using up/down switch
- Fixed feet
- Available width rage: 3 to 6 ft.
When installing the Cabinet on the hydraulic lift base stand, ensure that the hydraulic lines and the electrical cord are clear of any obstructions before installing the cabinet on the stand or operating the lift system.
Specifications
34.5 to 25.5 in. | |
66 Kg | |
39.7 in. | |
100.8 cm | |
115 V |
145 lb. | |
34.3 in. | |
87.1 cm | |
Electric Hydraulic Lift | |
ADA, NSF 49, UL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction