Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Mini Stoppering Chamber
GSA_VASDP

Catalog No. 10269825 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
FreeZone™ Freeze Dry System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
10-269-825 FreeZone™ Freeze Dry System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-269-825 Supplier Labconco™ Supplier No. 7569000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

FreeZone Freeze Dry System to attach Mini Stoppering Chamber via port

For lyophilizing samples in small serum vials with split stoppers

  • For use with a FreeZone™ Freeze Dry System
  • Accommodates 48 serum vials of 2 mL and 8 serum vials of 30 mL
  • After lyophilization, vials can be stoppered while still under vacuum
  • Stoppering travel distance: 0.5 in. (1.3 cm)
  • For vials ranging in height from 1.38 to 2.48 in. (3.5 to 6.3 cm)
  • Comes with tray for loading and unloading of samples

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Type Mini Stoppering Chamber
No. of Shelves 0
Material Acrylic
For Use With (Application) For lyophilizing samples in small serum vials with split stoppers
For Use With (Equipment) FreeZone™ Freeze Dry System
Includes 17 in. of clear pvc tubing
Width (English) 12 in.
Width (Metric) 30.4 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Weight (English) 15 lb.
Weight (Metric) 6.8 kg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.