Learn More
Labconco™ Positive Air Conversion Kit for Purifier™ HEPA Filter Glove Box
Change the built-in blower operation from standard negative pressure to positive pressure operation.
Supplier: Labconco™ 5441200
Description
This kit changes the built-in blower operation from standard negative pressure to positive pressure operation. The blower pushes instead of pulls HEPA/ULPA filtered air through the main chamber. Typical applications for positive pressure operation include handling materials that are highly sensitive to airborne particulate contaminants. The Positive Pressure Conversion Kit creates particulate cleanliness inside the box, measured to ISO 14644-1 test method, achieving ISO Class 3 conditions (1 particle 0.5 μm or larger per cubic foot of air per minute). Kit includes hose, two clamps and harness assembly. Not recommended for use with applications involving hazardous substances.Specifications
| Purifier HEPA Filter Clean Bench | |
| Conversion Kit | |
| Water service |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov