Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Positive Pressure Conversion Kit for Precise™ Glove Boxes
SDP

Catalog No. 16108401 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise HEPA Filtered Glove Box, Xpert Weigh Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16-108-401 Precise HEPA Filtered Glove Box, Xpert Weigh Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-108-401 Supplier Labconco™ Supplier No. 5236900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for Precise HEPA-filtered glove boxes, XPert weigh boxes

  • Contains components necessary to convert the built-in blower from negative pressure to positive pressure operation
  • For use w/Precise HEPA-filtered glove boxes, XPert weigh boxes

Specifications

For Use With (Equipment) Precise HEPA Filtered Glove Box, Xpert Weigh Box
Weight (English) 6 lb.
Weight (Metric) 2.7 Kg
Product Type Positive Pressure Conversion Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.