Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Combination Glove Boxes
SDP

Catalog No. 16200455 Shop All Labconco Corporation Products
Change view
Click to view available options
Certifications/Compliance:
CE
UL, ETL, CAN/CSA
Material:
Fiberglass
Stainless Steel
Filter Type:
HEPA
ULPA
Plug Type:
115V
230 V Plug
China/Australia Plug
Schuko Plug
UK Plug
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Material Filter Type Plug Type
16-200-455 UL, ETL, CAN/CSA Stainless Steel ULPA 230 V Plug
16-200-450 UL, ETL, CAN/CSA Fiberglass HEPA 230 V Plug
16-200-449 CE Fiberglass HEPA China/Australia Plug
16-200-448 CE Fiberglass HEPA Schuko Plug
16-200-447 CE Fiberglass HEPA UK Plug
16-200-446 UL, ETL, CAN/CSA Fiberglass HEPA 115V
16-200-451 UL, ETL, CAN/CSA Stainless Steel ULPA 115V
16-200-452 CE Stainless Steel ULPA UK Plug
16-200-453 CE Stainless Steel ULPA Schuko Plug
16-200-454 CE Stainless Steel ULPA China/Australia Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 16-200-455 Supplier Labconco™ Supplier No. 5080180
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Requires

Supporting base, vacuum pump for evacuating, inert gas for filling.

Can function as either a filtered box or a controlled atmosphere glove box

Opening two internal valves converts the box from controlled atmosphere mode to filtered mode. Ideal for laboratories that have multiple types of glove box applications or anticipate that their applications may change in the future.

Overall Features

  • Factory leak-tested—no detectable leaks greater than 1 × 10-6mL/sec.
  • LCD dual display—one shows pressure of both chambers, the other shows airflow (CFM), air changes/min. (ACM) and main chamber pressure (in. of water)
  • 0.38 in. thick laminated safety glass viewing window with 36.0W × 27.6 in.H (91 × 70cm) viewing area
  • Pair of neoprene gloves 0.015 in. thick, 30 in. long, size 9 3/4
  • Two 8 in. ID epoxy-coated aluminum, double-grooved glove ports with neoprene gaskets
  • Switches for light, blower and electrical receptacle
  • Two 25-watt fluorescent lights
  • Dry-powder epoxy-coated, 18 and 20 gauge steel exterior panels

Filtered Glove Box Features

  • Particulate containment <20 nanograms per cubic meter (naproxen sodium surrogate)
  • Fast “clean up” time—0.05 ppm average tracer gas levels or less detected during perimeter scan testing (surpasses ASHRAE 110 standards) after 5 minute dilution
  • Airborne particulate cleanliness exceeds ISO Class 4 conditions and achieves ISO Class 3 conditions
  • Inlet and outlet particulate (HEPA or ULPA) filters to prevent contaminants from entering or leaving the glove box chamber
  • Test port for challenging HEPA or ULPA filter integrity
  • True Bag-In/Bag-Out Filter Disposal System for safe filter removal and replacement
  • Built-in blower with speed control capable of airflow from 15 CFM to 85 CFM depending on voltage and frequency
  • Mutable, audible/visual low airflow/pressure alarm
Controlled Atmosphere Glove Box Features
  • Exceed Class 1 atmosphere containment conditions for oxygen permeation <1.67 ppm/min. per ISO 10648-2 test method
  • Four manual valves with 3/8 in. compression fittings—two on the main chamber and two on the transfer chamber
  • Automatic control of the main chamber pressure, main chamber evacuation and filling cycles, and transfer chamber evacuation and filling cycles
  • Regulation of vacuum/pressure between -5 and +5 inches of water gauge in the main chamber and between 0 and -29.5 inches of mercury in the transfer chamber
  • Mutable, audible/visual alarm that sounds when pressure/vacuum level is outside set limits
  • Three-position foot pedal (increase gas/fill, off, evacuate/decrease gas)
  • Factory-installed, rear-mounted, low profile stainless steel pressure relief bubbler at ±6 inches of water gauge
  • Two rear-mounted inlet and outlet 3/4 in. OD compression fittings for easy connection to optional gas purifier or accessory drying train

Certifications

ETL listed, UL 61010-1 and CAN/CSA C22.2 No. 1010.1 conformity (North American models); CE marked (International models)

Specifications

Product Type Glove Box
Filter Type ULPA
Lighting Type Fluorescent (2 x 25 W)
Material Stainless Steel
Amperage 10 A
Certifications/Compliance UL, ETL, CAN/CSA
Depth (English) 34.4 in.
Depth (Metric) 87.4 cm
Display LCD
For Use With (Application) Laboratories that have multiple types of glove box applications or anticipate that their applications may change in the future (atmosphere sensitive materials and hazardous materials)
For Use With (Equipment) AtmosPure™ Re-Gen gas purifier
Height (English) 45.7 in.
Height (Metric) 116.1 cm
Plug Type 230 V Plug
Ports 2
Voltage 100/115 V
Weight (English) 650 lb.
Weight (Metric) 295 kg
Width (English) 63.5 in.
Width (English) Interior 35.5 in.
Width (Metric) 161.3 cm
Width (Metric) Interior 90.17 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.