Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Double Stainless Steel Combination Glove Boxes
SDP

Catalog No. 16200459 Shop All Labconco Corporation Products
Change view
Click to view available options
Certifications/Compliance:
CE
UL, ETL, CAN/CSA
Plug Type:
China/Australia Plug
North America, 100/115 V
North America, 208/230 V
Schuko Plug
UK Plug
Voltage:
100/115 V
208/230 V
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Plug Type Voltage
16-200-459 CE China/Australia Plug 208/230 V
16-200-458 CE Schuko Plug 208/230 V
16-200-457 CE UK Plug 208/230 V
16-200-456 UL, ETL, CAN/CSA North America, 100/115 V 100/115 V
16-200-460 UL, ETL, CAN/CSA North America, 208/230 V 208/230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 16-200-459 Supplier Labconco™ Supplier No. 5080177
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Includes

Auto-Pressure Controller

Has the capability to function as either a filtered box or a controlled atmosphere glove box

  • Inner and outer pivoting transfer chamber doors, counterbalanced and equipped with quick-latches
  • Two rear-mounted inlet and outlet 0.75 in. OD compression fittings for connection to optional AtmosPure™ Re-Gen Gas Purifier or other accessory drying train
  • Interior right-hand side wall-mounted electrical receptacle (duplex on 100/115V models) with 10A maximum
  • Interior left-hand side wall-mounted pass-through with plug
  • Exceed Class 1 atmosphere containment conditions for oxygen permeation <1.67ppm/min per ISO 10648-2 test method
  • Centrally-mounted electrical receptacle
  • Type 304 stainless steel liner, Type 304 stainless steel inner and outer transfer chamber doors and Type 304 stainless steel front panel
  • Four fluorescent lights with two viewing windows and Four neoprene gloves and glove ports
  • Illumination exceeds 80 footcandles
  • Work with both atmosphere-sensitive and/or hazardous materials.

Specifications

Product Type Glove Box
Filter Type ULPA
Lighting Type Fluorescent (2 x 25 W)
Material Stainless Steel
Amperage 10 A
Certifications/Compliance CE
Depth (English) 34.4 in.
Depth (Metric) 87.4 cm
Display LCD
For Use With (Application) Laboratories that have multiple types of glove box applications or anticipate that their applications may change in the future (atmosphere sensitive materials and hazardous materials)
For Use With (Equipment) AtmosPure™ Re-Gen gas purifier
Height (English) 45.7 in.
Height (Metric) 116.1 cm
Plug Type China/Australia Plug
Ports 4
Voltage 208/230 V
Weight (English) 1300 lb.
Weight (Metric) 590 kg
Width (English) 108.9 in.
Width (English) Interior 81.8 in.
Width (Metric) 276.6 cm
Width (Metric) Interior 207.77 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.