Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Downdraft Powder Station
SDP

Catalog No. 09681306 Shop All Labconco Corporation Products
Change view
Click to view available options
Voltage:
115 V
230 V
Width (Metric):
60.9 cm
91.4 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Width (Metric)
09-681-306 230 V 91.4 cm
09-681-302 230 V 60.9 cm
09-681-301 115 V 60.9 cm
09-681-305 115 V 91.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Does not provide protection from biohazardous aerosols or gaseous contaminants. Contact your Fisher Customer Service Representative for information on biosafety cabinets and fume hoods if your application involves these contaminants.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 09-681-306 Supplier Labconco™ Supplier No. 3410021
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Does not provide protection from biohazardous aerosols or gaseous contaminants. Contact your Fisher Customer Service Representative for information on biosafety cabinets and fume hoods if your application involves these contaminants.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

The Labconco Protector™ Downdraft Powder Station keeps users safe by containing powders during fingerprint processing.

Open-sided, open-top workstation provides unrestricted operator movement and accommodates oversized evidence.

  • Powder-coated steel housing and shelf
  • Type 304 stainless steel work surface with Flow-Zone™ perforations
  • Built-in blower with motorized, backward curved fan and switch
  • With removable powder catch pan
  • Exhaust prefilter of pleated paper with MERV 8 rating
  • Exhaust HEPA filter, 99.99% efficient on particles 0.3 micron
  • Red, filter system alarm light alerts operator to low airflow conditions
  • Comes with 6 ft. (15cm) power cord with plug    
  • Available in 2 ft. (0.6m) and 3 ft. (0.9m) widths and Domestic electrical standards

An optional Transition Connector mounts between two downdraft powder hoods for a secure fit and continuous work surface.

Compliance

ETL listed (Models 3410000 and 3410001).

Specifications

Product Type Downdraft Powder Station
Coating Epoxy Powder
Filter Type HEPA
Material Stainless Steel
Amperage 1 A
Depth (English) 25.7 in.
Depth (Metric) 65.3 cm
Height (English) 18.75 in.
Height (Metric) 47.6 cm
Voltage 230 V
Weight (English) 87 lb.
Weight (Metric) 39 kg
Width (English) 36 in.
Width (Metric) 91.4 cm
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.