Learn More
Labconco™ Protector Glove Box Accessory Kits
SDP
Catalog No. 16200386 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Add-A-Valve Kit for Main Chamber
Pressure Relief Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16-200-386 Add-A-Valve Kit for Main Chamber
16-200-387 Pressure Relief Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-200-386 Supplier Labconco™ Supplier No. 5442700
Add to Cart
Add to Cart
Kits provided added functionality options for Protector glove boxes
Add-A-Valve Kits include two manual valves, tubes, nuts and instructions for adding valves to the glove box for evacuating with vacuum, filling with inert gas, connecting to a drying train, or connecting to filtration components. Manual valves mount in predrilled holes behind the label located below the control panel. For use with Protector Combination and Protector Filtered Glove Boxes.
Specifications
For Use With (Equipment) Protector Glove Boxes
Includes 2 Manual Valves, Nuts, Instructions
Product Type Add-A-Valve Kit for Main Chamber