Learn More
Labconco™ Protector Glove Box Accessory Kits
SDP
Catalog No. 16200387 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Add-A-Valve Kit for Main Chamber
Pressure Relief Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16-200-387 Pressure Relief Kit
16-200-386 Add-A-Valve Kit for Main Chamber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-200-387 Supplier Labconco™ Supplier No. 5441403
Add to Cart
Add to Cart
Kits provided added functionality options for Protector glove boxes
Kit Components
  • 5 in. diameter round HEPA filter (99.9% efficient on 0.3 micron particulates)
  • Plastic hose barbs (2)
  • 10 ft. of 3/4 in. ID neoprent tubing
Specifications
For Use With (Equipment) Protector Glove Boxes
Includes HEPA filter, hose barb connectors and 10 ft. of 0.75 in. ID neoprene tubing for attachment to the pressure relief bubbler exhaust
Product Type Pressure Relief Kit