Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Work Surface Accessory, Oval Polypropylene Cupsink
SDP

Catalog No. 1630615 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Flat or SpillStopper™ work surface with cutout
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16-306-15 Flat or SpillStopper™ work surface with cutout
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-306-15 Supplier Labconco™ Supplier No. 4005200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easily remove waste from your laboratory worktable with the Labconco Oval Cupsink.

  • Works with flat or Labconco SpillStopper worksurfaces with cutout
  • Polypropylene
  • Oval
  • Weighs only 2 lbs.

Specifications

Color Silver
Material Polypropylene
Type Cupsink Cover
For Use With (Equipment) Flat or SpillStopper™ work surface with cutout
Shape Oval
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.