Learn More
Labconco™ Protector XVS™ PVC Female Duct Coupling for Ventilation Stations
SDP
Catalog No. 1630412 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector VS Ventilation Station
Catalog No. For Use With (Equipment)
16-304-12 Protector VS Ventilation Station
1 options
Catalog No. 16-304-12 Supplier Labconco™ Supplier No. 4708900
Add to Cart
Add to Cart
PVC Female Duct Coupling for Ventilation Stations
  • For use with Labconco Protector XVS Ventilation Stations
Specifications
For Use With (Equipment) Protector VS Ventilation Station
Weight (English) 4 lb.
Weight (Metric) 1.8 Kg
Includes Female Duct Coupling
Dimensions 6 in. dia. round PVC duct
Material PVC
For Use With (Application) Ducting