Learn More
Labconco™ Protector XVS™ Zero–Pressure PVC Weathercap for Ventilation Stations
SDP
Catalog No. 1630414 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector VS Ventilation Station
Catalog No. For Use With (Equipment)
16-304-14 Protector VS Ventilation Station
1 options
Catalog No. 16-304-14 Supplier Labconco™ Supplier No. 4722200
Add to Cart
Add to Cart
Pressure PVC Weathercap for Ventilation Stations
  • For use with Labconco Protector XVS Ventilation Stations
  • PVC zero-pressure weathercap connects directly to duct for vertical discharge of effluent air and dispersion away from building
  • Cap keeps out rain and snow but adds little static pressure to exhaust system
Specifications
For Use With (Equipment) Protector VS Ventilation Station
Height (English) 36 in.
Weight (English) 20 lb.
Weight (Metric) 9.1 Kg
Includes Weathercap
Dimensions 6 in. dia. round PVC duct
Material PVC
For Use With (Application) Ducting