Learn More
Labconco™ PuriCare™ Open Access Station
Labconco PuriCare™ Open Access Station allows trouble-free transfer of animals from dirty to clean cages.
Manufacturer:  Labconco™ 3820000
 View more versions of this product
Catalog No. 10-269-131
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
12Amp
24.13 in.
61.29cm
115V 60Hz
Cage changing and animal transfer
14 in.
35.56cm
60Hz
Two 32w fluorescent
115V
600 lb.
48.5 in.
123.19cm
76.6 to 88.6 in.
ADA, ETL, UL
30 in.
76cm
2 HEPA
78.31 to 84.31 in.
198.9 to 214.1cm
198 to 220cm
28 ft. orange, retractable, 3-wire cord and plug, 2 push and pull bars
Epoxy Coated Steel Exterior and Type 304 Stainless Steel Work Surface
3 Years
272kg
58.4 in.
148cm