Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Cell Logic+™ Class II A2 Biosafety Cabinets, 4ft. Width, 9in. Opening, International Model
SDP

Catalog No. p-4588057 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Procedure Station, UV Light, Service Fixtures, Pass-through
Scope-ready Package, Service Fixtures, Pass-through, Base Stand
Scope-ready Package, Temp-zone Package, Service Fixtures, Pass-through
Temp-zone Package, UV Light, Service Fixtures, Pass-through
Temp-zone Package, UV Light, Service Fixtures, Pass-through, Base Stand
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
33-249-1160 Scope-ready Package, Service Fixtures, Pass-through, Base Stand
32-249-1060 Temp-zone Package, UV Light, Service Fixtures, Pass-through
32-249-1160 Temp-zone Package, UV Light, Service Fixtures, Pass-through, Base Stand
34-249-1060 Scope-ready Package, Temp-zone Package, Service Fixtures, Pass-through
34-249-1160 Scope-ready Package, Temp-zone Package, Service Fixtures, Pass-through
33-249-1060 Procedure Station, UV Light, Service Fixtures, Pass-through
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 33-249-1160 Supplier Labconco™ Supplier No. 332491160

missing translation for 'validMainframeMsg'

Biohazard cabinets with advanced ergonomic design excel in user comfort and functionality

MyLogic™ operating system provides intuitive programming, color-coded menus, status messages and alarms with diagnostic troubleshooting — all using full sentences.

  • Same ECM blower technology as Purifier Logic™; uses Constant Airflow Profile™ (CAP) to maintain precise volume of air as filters load
  • As an additional safety feature, cabinet will continuously display HEPA filter life remaining and cabinet status
  • Provides personnel, product and environmental protection; suitable for work in Biosafety Levels 1, 2 and 3
  • Discharges HEPA-filtered exhaust air directly into lab, or into an exhaust system through optional Canopy Connection
  • Equipped to meet the specific needs of the cell culture industry
  • Available with optional Scope-Ready™ package, Temp-Zone™ package or both
  • Scope-Ready package allows the users microscope to be integrated into the biosafety cabinet
  • Temp-Zone package includes a temperature controlled area on the work surface
Inclination Technology™ Suite includes:
  • 10° angled sash
  • Patented curved air grille with built-in armrest
  • Fully accessible controls and eye-level mounted color display with MyLogic operating system
  • Flush-mounted, slowly self-closing electrical outlet covers
  • ECM is quieter and cooler than comparable PSC motors
  • Large interior work area

Some cabinets include non-welded base stands. For cabinets that do not include the base stand, one is required and is sold separately.

Specifications

Certifications/Compliance CE, ETL, ADA, NSF 49, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel
Noise Level <63 dBA
Plug Type 230 V EN 12469
Height (English) 89.3 to 95.3 in.
Sash Opening (Metric) 22.6 cm
Display LCD
Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
Includes Scope-ready Package, Service Fixtures, Pass-through, Base Stand
Lighting Fluorescent
Height (Metric) 226.8 to 242 cm
Width (English) 54.3 in.
Width (Metric) 137.9 cm
Frequency 50/60 Hz
Height (English) Viewing 27 in.
Depth (English) 31.2 in.
Depth (Metric) 79.2 cm
Weight (English) 670 lb.
Weight (Metric) 303.9 Kg
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Amperage 6 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.