Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Purifier™ Horizontal Clean Benches, 8 ft.

Protects work from particulate contamination
Supplier: Labconco™ 3818521
Description
- Horizontal laminar flow
- Double-gasket, negative pressure HEPA filter leak protection
- ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
- Disposable prefilters (2 each on 3 ft. and 4 ft. models; 3 each on 6 ft. models; 4 each on 8 ft. models)
- Variable speed built-in motor/blower(s) with vibration isolation pads
- Differential pressure gauge
- Type 304 brushed stainless-steel work surface
- Tempered safety glass side panels with utility ports with iris openings
- UV-resistant, dry powder epoxy-coated steel exterior
- Front-mounted light and blower switches
Warranty and Services
One year warranty on parts and service
Specifications
Fluorescent | |
66 cm | |
122.9 cm | |
Domestic | |
388 kg | |
254 cm |
26 in. | |
48.4 in. | |
60 Hz | |
855 lb. | |
100 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction