Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Logic+™ Class II A2 Biosafety Cabinet, 6 ft. Width, 8 in. Opening, International Model
SDP

Catalog No. 302681040 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Base Stand
Base Stand, UV Light, Service Fixtures, Pass-through
Cabinet Only
UV Light, Service Fixtures, Pass-through
Lighting:
Fluorescent
Fluorescent, UV
Voltage:
100 V
230 V
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Lighting Voltage
30-268-1040 UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-0040 Cabinet Only Fluorescent 230 V
30-268-0030 Cabinet Only Fluorescent 230 V
30-268-0020 Cabinet Only Fluorescent 100 V
30-268-0050 Cabinet Only Fluorescent 230 V
30-268-0070 Cabinet Only Fluorescent 230 V
30-268-0150 Base Stand Fluorescent 230 V
30-268-0140 Base Stand Fluorescent 230 V
30-268-0130 Base Stand Fluorescent 230 V
30-268-0120 Base Stand Fluorescent 100 V
30-268-0170 Base Stand Fluorescent 230 V
30-268-1020 UV Light, Service Fixtures, Pass-through Fluorescent, UV 100 V
30-268-1050 UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-1030 UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-1070 UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-1120 Base Stand, UV Light, Service Fixtures, Pass-through Fluorescent, UV 100 V
30-268-1130 Base Stand, UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-1140 Base Stand, UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-1150 Base Stand, UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
30-268-1170 Base Stand, UV Light, Service Fixtures, Pass-through Fluorescent, UV 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 30-268-1040 Supplier Labconco™ Supplier No. 302681040

missing translation for 'validMainframeMsg'

Labconco Purifier™ Logic+™ Class II A2 Biosafety Cabinets with advanced ergonomic design excel in user comfort and functionality.

These cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through an optional canopy connection.

  • Electronically Commutated Motor (ECM)
  • Built-in interval or elapsed timer for experiment monitoring, fluorescent light or UV light control (on models with UV light)
  • LCD information center with “Filter Life Remaining” bar graph, status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
  • Electronic security lock (optional activation) that requires code to operate the cabinet
  • Two electrical duplex receptacles, (single outlets on 230 volt models), located one on each side, with ground fault interruption (115 volt models)
  • Stainless steel inlet grille with Reserve-Air Secondary Airflow Slots
  • Interior-mounted, line-of-sight, full color LCD information center with easy-to-understand MyLogic OS
  • Flush-mounted stainless steel electrical receptacle covers with dampened hinges
  • Removable, seamless type 304 stainless steel, dished work surface with lift out knobs
  • ADA-compliant touchpad control on right-hand side post for manual activation of blower, light(s), timer, electrical receptacles, audible alarm mute and menu selection

  • Fully-closing, clear 6.35mm (1/4 in.) tempered safety glass sash
  • 54cm (21.7 in.) maximum sash opening height and 69cm (27.0 in.) viewing height
  • Nominal inflow velocity of 105 feet per minute (fpm) (0.5 m/sec)
  • Nominal downflow velocity of 55 fpm (0.3 m/sec)
  • Approximately 70% air recirculation
  • Powder-coated steel exterior
  • 3m (10 ft.) power cord with plug

For work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates

Certifications

NSF Standard Number 49 and ETL listed.

Compliance

CE conformity marking (230 volt models). Class 5 conditions per ISO 14644-1 and 2 (formerly Class 100).

Specifications

Certifications/Compliance ADA, CE, NSF 49
Chamber Material Stainless Steel
Noise Level <63 dBA
Height (English) 61.7 in.
Sash Opening (Metric) 20.3 cm
Display LCD
Housing Material Powder Coated Steel Exterior and Stainless Steel Interior
Includes UV Light, Service Fixtures, Pass-through
Lighting Fluorescent, UV
Height (Metric) 156.7 cm
Width (English) 78.3 in.
Width (Metric) 198.8 cm
Height (English) Viewing 27 in.
Depth (English) 31.2 in.
Depth (Metric) 79.2 cm
Weight (English) 805 lb.
Weight (Metric) 365.1 Kg
Voltage 230 V
Amperage 8 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.