Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Vertical Clean Bench
SDP

Catalog No. 16108251 Shop All Labconco Corporation Products
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
Voltage:
115 V
230 V
Plug Type:
No Plug
North America Plug
Includes:
Airflow Monitor
Bench Only
UV, Panel, Timer
UV, Panel, Timer, Airflow Monitor
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Voltage Plug Type Includes
16-108-251 24 in. 230 V No Plug UV, Panel, Timer, Airflow Monitor
16-108-243 24 in. 115 V North America Plug Bench Only
16-108-248 24 in. 230 V No Plug Bench Only
39-702-03 24 in. 115 V North America Plug Airflow Monitor
16-108-250 24 in. 230 V No Plug Airflow Monitor
16-108-249 24 in. 230 V No Plug UV, Panel, Timer
16-108-252 36 in. 230 V North America Plug Bench Only
16-108-256 36 in. 230 V No Plug Bench Only
16-108-258 36 in. 230 V No Plug Airflow Monitor
16-108-253 36 in. 230 V North America Plug UV, Panel, Timer
16-108-257 36 in. 230 V No Plug UV, Panel, Timer
16-108-259 36 in. 230 V No Plug UV, Panel, Timer, Airflow Monitor
16-108-264 48 in. 230 V No Plug Bench Only
16-108-266 48 in. 230 V No Plug Airflow Monitor
16-108-261 48 in. 115 V North America Plug UV, Panel, Timer
16-108-265 48 in. 230 V No Plug UV, Panel, Timer
16-108-263 48 in. 115 V North America Plug UV, Panel, Timer, Airflow Monitor
16-108-267 48 in. 230 V No Plug UV, Panel, Timer, Airflow Monitor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 16-108-251 Supplier Labconco™ Supplier No. 3970224
Only null left
Add to Cart
Add to Cart
Request a Quote
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Requires

Work surface and base stand (sold separately).

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.

  • Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
  • Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
  • Intrinsically-sound, negative pressure design
  • Minihelic pressure gauge and disposable prefilter
  • UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
  • UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
  • Light and blower switches
  • Air diffuser and 23.8cm (9.4 in.) high sash opening
  • Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
  • Two utility ports with iris openings and plugs
  • ETL Listing (115 volt, 60 Hz models)

Order Info

Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.

Warranty and Services

One year parts and service

Specifications

Product Type Clean Bench
Coating Powder
Filter Type HEPA
Lighting Type Fluorescent
Material Powder-coated Aluminum and Steel
Amperage 2 A
Certifications/Compliance CAN/CSA C22.2, UL, CE
Color Glacier White, Gray
Depth (English) 28.6 in.
Depth (English) Interior 25.3 in.
Depth (Metric) 72.64 cm
Depth (Metric) Interior 64.3 cm
For Use With (Application) Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
Height (English) 36.8 in.
Height (English) Interior 22.7 in.
Height (Metric) 93.47 cm
Height (Metric) Interior 57.7 cm
Hertz 50/60 Hz
Includes UV, Panel, Timer, Airflow Monitor
IP Rating None
Plug Type No Plug
Ports Utility
Voltage 230 V
Weight (English) 125 lb.
Weight (Metric) 57 kg
Width (English) 24 in.
Width (English) Interior 22.8 in.
Width (Metric) 60.96 cm
Width (Metric) Interior 57.9 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.