Learn More
Labconco™ Purifier™ Vertical Clean Bench

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.
Supplier: Labconco™ 3970320
Alert:
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Requires : Work surface and base stand (sold separately).
Description
- Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
- Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
- ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
- Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
- Intrinsically-sound, negative pressure design
- Minihelic pressure gauge and disposable prefilter
- UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
- UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
- Light and blower switches
- Air diffuser and 23.8cm (9.4 in.) high sash opening
- Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
- Two utility ports with iris openings and plugs
- ETL Listing (115 volt, 60 Hz models)
Order Info
Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.
Warranty and Services
One year parts and service
Specifications
| Clean Bench | |
| HEPA | |
| Powder-coated Aluminum and Steel | |
| CAN/CSA C22.2, UL, CE | |
| 28.6 in. | |
| 72.64 cm | |
| Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation | |
| 22.7 in. | |
| 57.7 cm | |
| Bench Only | |
| No Plug | |
| 230 V | |
| 73 kg | |
| 34.8 in. | |
| 88.4 cm |
| Powder | |
| Fluorescent | |
| 3 A | |
| Glacier White, Gray | |
| 25.3 in. | |
| 64.3 cm | |
| 36.8 in. | |
| 93.47 cm | |
| 50/60 Hz | |
| None | |
| Utility | |
| 160 lb. | |
| 36 in. | |
| 91.44 cm |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.