Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Vertical Clean Bench
SDP

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.

Supplier:  Labconco™ 3970321

 View more versions of this product

Catalog No. 16-108-257


Only null left
Add to Cart
Request a Quote

Alert:

Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).

Alert:

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.


Requires : Work surface and base stand (sold separately).

Description

Description

  • Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
  • Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
  • Intrinsically-sound, negative pressure design
  • Minihelic pressure gauge and disposable prefilter
  • UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
  • UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
  • Light and blower switches
  • Air diffuser and 23.8cm (9.4 in.) high sash opening
  • Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
  • Two utility ports with iris openings and plugs
  • ETL Listing (115 volt, 60 Hz models)

Order Info

Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.

Warranty and Services

One year parts and service

Specifications

Specifications

Clean Bench
HEPA
Powder-coated Aluminum and Steel
CAN/CSA C22.2, UL, CE
28.6 in.
72.64 cm
Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
22.7 in.
57.7 cm
UV light, protection panel and timer
No Plug
230 V
73 kg
34.8 in.
88.4 cm
Powder
Fluorescent
3 A
Glacier White, Gray
25.3 in.
64.3 cm
36.8 in.
93.47 cm
50/60 Hz
None
Utility
160 lb.
36 in.
91.44 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.