Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Purifier™ Vertical Clean Bench
GSA_VASDP

Catalog No. 3970203 Shop All Labconco Corporation Products
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
Voltage:
115 V
230 V
Plug Type:
No Plug
North America Plug
Includes:
Airflow Monitor
Bench Only
UV, Panel, Timer
UV, Panel, Timer, Airflow Monitor
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Voltage Plug Type Includes
39-702-03 24 in. 115 V North America Plug Airflow Monitor
16-108-243 24 in. 115 V North America Plug Bench Only
16-108-248 24 in. 230 V No Plug Bench Only
16-108-250 24 in. 230 V No Plug Airflow Monitor
16-108-249 24 in. 230 V No Plug UV, Panel, Timer
16-108-251 24 in. 230 V No Plug UV, Panel, Timer, Airflow Monitor
16-108-256 36 in. 230 V No Plug Bench Only
16-108-258 36 in. 230 V No Plug Airflow Monitor
16-108-253 36 in. 230 V North America Plug UV, Panel, Timer
16-108-257 36 in. 230 V No Plug UV, Panel, Timer
16-108-259 36 in. 230 V No Plug UV, Panel, Timer, Airflow Monitor
16-108-264 48 in. 230 V No Plug Bench Only
16-108-266 48 in. 230 V No Plug Airflow Monitor
16-108-265 48 in. 230 V No Plug UV, Panel, Timer
16-108-263 48 in. 115 V North America Plug UV, Panel, Timer, Airflow Monitor
16-108-267 48 in. 230 V No Plug UV, Panel, Timer, Airflow Monitor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).
Catalog No. 39-702-03 Supplier Labconco™ Supplier No. 3970203
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Purifier Vertical Clean Benches do not provide user protection. Biohazardous materials, toxins and radionuclides should be handled in Class I or Class II biological safety cabinets. (See Purifier Class I Safety Enclosures and Delta Series Cabinets).

Requires

Work surface and base stand (sold separately).

Protect work from particulate contamination by choosing a Purifier™ Vertical Clean Bench.

  • Nominal downflow velocity of 45-65 fpm and aerodynamic air foil
  • Supply HEPA filter, 99.99% efficient on particles 0.3 Micron
  • ISO Class 5 air (formerly Class 100 per Federal Standard 209E)
  • Variable speed built-in blower with motorized impeller and solid state control rated for 10 A
  • Intrinsically-sound, negative pressure design
  • Minihelic pressure gauge and disposable prefilter
  • UV-resistant, glacier white and gray, powder-coated aluminum and steel construction
  • UV-opaque and resistant 6.3mm (0.25 in.) thick tempered safety glass sash and sides
  • Light and blower switches
  • Air diffuser and 23.8cm (9.4 in.) high sash opening
  • Ergonomic 10° angled and hinged sash that pivots upward, locks to a loading height of 50.8cm (20 in.) and has wiping seals to promote ISO Class 5 conditions
  • Two utility ports with iris openings and plugs
  • ETL Listing (115 volt, 60 Hz models)

Order Info

Vertical Clean Benches can be ordered with bifold doors; specify when ordering. Guardian™ Airflow Monitor is offered as factory-installed option on all benches; specify when ordering.

Warranty and Services

One year parts and service

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Clean Bench
Coating Powder
Filter Type HEPA
Lighting Type Fluorescent
Material Powder-coated Aluminum and Steel
Amperage 5 A
Certifications/Compliance CAN/CSA C22.2, UL, CE
Color Glacier White, Gray
Depth (English) 28.6 in.
Depth (English) Interior 25.3 in.
Depth (Metric) 72.64 cm
Depth (Metric) Interior 64.3 cm
For Use With (Application) Plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation
Height (English) 36.8 in.
Height (English) Interior 22.7 in.
Height (Metric) 93.47 cm
Height (Metric) Interior 57.7 cm
Includes Airflow Monitor
IP Rating None
Plug Type North America Plug
Ports Utility
Voltage 115 V
Weight (English) 125 lb.
Weight (Metric) 57 kg
Width (English) 24 in.
Width (English) Interior 22.8 in.
Width (Metric) 60.96 cm
Width (Metric) Interior 57.9 cm
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.