Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Recirculation Kit for Precise™ Glove Boxes
SDP

Catalog No. 16108402 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise HEPA Filtered Glove Box, Xpert Weigh Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
16-108-402 Precise HEPA Filtered Glove Box, Xpert Weigh Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-108-402 Supplier Labconco™ Supplier No. 5236500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for Precise Glove Boxes

  • Contains components necessary to connect the built-in blower exhaust to the inlet HEPA filter for recirculation of air
  • Useful for dry atmosphere operations

Specifications

For Use With (Equipment) Precise HEPA Filtered Glove Box, Xpert Weigh Box
Weight (English) 5.5 lb.
Weight (Metric) 2.5 Kg
Product Type Recirculation Kit
For Use With (Application) Dry Atmosphere Operations
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.