Learn More
Labconco™ Remote Blower for Protector™ Workstations
GSA_VASDP
Catalog No. 1630745 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Work Station
Catalog No. For Use With (Equipment)
16-307-45 Protector Work Station
1 options
Catalog No. 16-307-45 Supplier Labconco™ Supplier No. 7053500
Add to Cart
Add to Cart
Remote Blower, for Labconco Protector Work Station without Blower.
Remote Blower, for Protector Work Station without Blower. Sized to meet the air volume requirements of the Protector Work Station 6941003. It has a 1/4 hp direct drive motor capable of overcoming external static pressure loss of .87 in. H2O at 167 CFM (50 fpm face velocity). Housing and wheel are corrosion-resistant enamel-coated steel. Discharge direction is adjustable to accommodate unusual blower locations. Blower inlet is 6 in. ID. Outlet dimension is 4.25 in. x 7.38 in. OD.
†External static pressure loss at 50fpm face velocity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Protector Work Station
Weight (English) 35 lb.
Weight (Metric) 15.9 Kg
Includes Remote blower
Voltage 115 V
For Use With (Application) Fumes
WARNING: Cancer - www.P65Warnings.ca.gov