Learn More
Labconco™ Replacement Bag for Hepa Filter Cell
SDP
Replacement bag for filters in XPert and RXPert Balance and Bulk Powder Enclosures and XPert Nano Enclosures.
Supplier:  Labconco™ 3776002
Catalog No. 10-304-61
Only null left
Add to Cart
Description
Description
Replacement Bag for Bag-in/Bag-out HEPA Filter for use with Labconco FilterMate Portable Exhausters.
Specifications
Specifications
HEPA
1 lb.
Filter Accessory
HEPA filter cell, Protector™ demonstration hood, FilterMate™ portable exhauster
0.5 Kg
Bag-in/bag-out HEPA filter to contain particulate matter during filter changing
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.