Learn More
Labconco™ Work Surface Accessory, PVC Trap
SDP
Catalog No. 16108234 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Coupling Nut
Catalog No. Includes
16-108-234 Coupling Nut
1 options
Catalog No. 16-108-234 Supplier Labconco™ Supplier No. 1432600
Add to Cart
Add to Cart
Customize your SpillStopper worksurface with a PVC Trap, designed to prevent backflow and odors from escaping.
  • For use with SpillStopper or other flat work surfaces outfitted with a cupsink
  • 1.5 in. diameter drum with coupling nut
  • Shipping weight: 5kg (10 lbs.)
Specifications
For Use With (Equipment) SpillStopper or other flat worksurfaces outfitted with cupsinks
Includes Coupling Nut
Product Type 1.5 in. Drum P-Trap
Material PVC
WARNING: Cancer - www.P65Warnings.ca.gov