Learn More
Labconco™ Retaining Bands
SDP
Catalog No. 10265174 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Natural Rubber, Latex
Catalog No. Material
10-265-174 Natural Rubber, Latex
1 options
Catalog No. 10-265-174 Supplier Labconco™ Supplier No. 3981200
Add to Cart
Add to Cart
Used to secure Safety Sleeve and inner source container bag to Retaining Ring
  • Used with XPert Bulk Powder Filtered Systems, Stations and Enclosures
  • Fits all sizes of Retaining Rings
  • Disposable or reusable after cleaning
Specifications
Material Natural Rubber, Latex
Weight (English) 1 lb.
Weight (Metric) 0.5 kg