Learn More
Labconco™ Rotors for CentriVap™ Systems
GSA_VASDP
For use with Labconco CentriVap Systems
$685.50 - $2820.00
Specifications
Material Anodized Aluminum
For Use With (Equipment) CentriVap™ DNA system
View More Specs
Products 6
Catalog Number Mfr. No. Holds Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Holds Weight (English) Weight (Metric) Price Quantity & Availability  
16-317-50
GSA_VASDP
View Documents
Labconco™
7450700
132 x 1.5 to 2 mL Microcentrifuge Tube 2 lb. 0.9 kg
Each for $685.50
Only null left
Add to Cart
16-317-53
GSA_VASDP
View Documents
Labconco™
7461900
4 Standard or 2 Deep-well Plates 3 lb. 1.4 kg
Each for $2,280.00
Only null left
Add to Cart
16-317-54
GSA_VASDP
View Documents
Labconco™
7462900
72 x 0.5 mL Microcentrifuge Tube or 60 x 1.5 to 2 mL Microcentrifuge Tube 2 lb. 0.9 kg
Each for $703.00
Only null left
Add to Cart
16-317-62
GSA_VASDP
View Documents
Labconco™
7455100
40 x 1.5 to 2 mL Microcentrifuge Tubes or 100 x 5 to 6 mL Tubes 3 lb. 1.4 kg
Each for $2,530.00
Only null left
Add to Cart
16-317-63
GSA_VASDP
View Documents
Labconco™
7455200
32 x 40 mL to 50 mL Microcentrifuge Tubes 3 lb. 1.4 kg
Each for $2,350.00
Only null left
Add to Cart
16-317-64
GSA_VASDP
View Documents
Labconco™
7455000
44 x 12 to 17 mL and 24 x 16 to 17 mL Tubes 4 lb. 1.8 kg
Each for $2,820.00
Only null left
Add to Cart
Description
Description
For use with Labconco CentriVap Systems
  • Anodized aluminum rotors hold tubes at angle to maximize surface area
  • Two hole sizes on rotors 16-317-54 and 16-317-62 to -64 accommodate tubes with different O.D.
  • Maximum tube capacity for a given O.D. may vary with overall tube height
  • CentriVap DNA Systems and Proteomic come with DNA Rotor (16-317-54); Acid Systems come with Acid-Resistant 12-13 mm Rotor (16-108-450)
To prevent loss of sample during centrifugation, fill tubes only 0.33 to 0.5 full.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Anodized Aluminum
CentriVap™ DNA system
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.