Learn More
Labconco™ Sash-Glider Kit
SDP
Catalog No. 09681310 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Sash-glider Kit
Catalog No. Product Type
09-681-310 Sash-glider Kit
1 options
Catalog No. 09-681-310 Supplier Labconco™ Supplier No. 3408500
Add to Cart
Add to Cart
Includes
Magnetic outside and inside blades, 2 microfiber sheets and 10 disposable white scrubbing strips
Used for cleaning both sides of biological safety cabinet's sash while it remains closed, to avoid contamination risks
  • Applicable for: PuriCare Procedure Stations (2013 and newer), Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets
Specifications
Weight (English) 5 lb.
Weight (Metric) 2 Kg
Includes Magnetic Outside and Inside Blades, 2 Microfiber Sheets and 10 Disposable White Scrubbing Strips
Product Type Sash-glider Kit