Learn More
Labconco™ Sash Stop Kit
SDP
Catalog No. 09039052 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Premier Hood, Protector XL Benchtop Hood
Protector Premier Hood, Protector XStream Hood
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09-039-052 Protector Premier Hood, Protector XL Benchtop Hood
10-369-203 Protector Premier Hood, Protector XStream Hood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-039-052 Supplier Labconco™ Supplier No. 9724500P
Add to Cart
Add to Cart
Sash stop restricts how far a vertical-rising sash may be opened.
  • For use with older models of Protector Premier, XL Benchtop, Stainless Steel Radioisotope and Stainless Steel Perchloric Acid Fume Hoods made before 2012.
  • Restricts how far a vertical-rising sash may be opened.
  • Plastic device may be easily field installed on the fixture panel of a Protector Hood, which have been factory-prepared to accommodate a sash stop at the 60% open position (18 in. to work surface).
  • Alternate sash positions may be field drilled. Each Sash Stop Kit includes components for one sash.
  • 1 kit required for 3 ft. to 8 ft. Premier, XL Benchtop, XStream, PVC, Stainless Steel and XL Floor-Mounted Fume Hoods with vertical-rising sashes.
  • 2 kits required for 10 ft., 12 ft. XL Benchtop with dual sash, Pass-Through Fume Hoods.
Specifications
For Use With (Equipment) Protector Premier Hood, Protector XL Benchtop Hood
Weight (English) 0.5 lb.
Weight (Metric) 0.2 Kg
Product Type Sash Stop