Learn More
Labconco™ Argon Standard Service Fixture Kit
SDP
Catalog No. 09039043 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector, XStream, XL, XLE Laboratory Hood
Catalog No. For Use With (Equipment)
09-039-043 Protector, XStream, XL, XLE Laboratory Hood
1 options
This item is not returnable. View return policy
Catalog No. 09-039-043 Supplier Labconco™ Supplier No. 9808800
Add to Cart
Add to Cart
This item is not returnable. View return policy
Kit includes copper tubing, brass valve and gray knob/connector.
Kit includes copper tubing, brass valve and gray knob/connector.
  • Max. flow rate of 20.2 CFM.
  • Max. working pressure of 40 psi.
  • Kits mount on the right or left side of any Protector Premier, XStream, XL, XLE or PVC Hood.
  • Each kit includes one remotely-controlled service fixture with valve and 0.25 in. diameter tubing, color-coded fixture knob and color-coded hose connector.
  • Specifications
    For Use With (Equipment) Protector, XStream, XL, XLE Laboratory Hood
    Weight (English) 4 lb.
    Weight (Metric) 1.8 Kg
    Includes 0.25 in. dia. Copper Tubing, Brass Valve and Green Knob, Connector
    Product Type Argon (ARG) Standard Service Fixture KitArgon (ARG) Standard Service Fixture Kit