Learn More
Labconco™ Deionized/Distilled Water Standard Service Fixture Kit
SDP
Catalog No. 09039045 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector, XStream, XL, XLE Laboratory Hood
Catalog No. For Use With (Equipment)
09-039-045 Protector, XStream, XL, XLE Laboratory Hood
1 options
This item is not returnable. View return policy
Catalog No. 09-039-045 Supplier Labconco™ Supplier No. 9809100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Kit includes stainless steel tubing, nickel-plated & stainless steel valve and white knob/connector.
Kit includes stainless steel tubing, nickel-plated & stainless steel valve and white knob/connector.
  • Max. flow rate of 3.5 GPM (13.2 LPM).
  • Max. working pressure of 40 psi.
  • Kits mount on the right or left side of any Protector Premier, XStream, XL, XLE or PVC Hood.
  • Each kit includes one remotely-controlled service fixture with valve and 0.25 in. diameter tubing, color-coded fixture knob and color-coded hose connector.
  • Specifications
    For Use With (Equipment) Protector, XStream, XL, XLE Laboratory Hood
    Weight (English) 4 lb.
    Weight (Metric) 1.8 Kg
    Includes 0.25 in. dia. Stainless Steel Tubing, Nickel-plated, Stainless Steel Valve and White Knob, Connector
    Product Type Deionized, Distilled Water (DI) Standard Service Fixture Kit