Learn More
Labconco™ SpillStopper™ Work Surface for 6ft. Protector™XStream
SDP
Catalog No. 10369195 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Left Rear
Left Rear, Right Rear
Left Side
Left Side, Right Rear
No Cutouts
Right Rear
Right Side
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
10-369-195 Left Side, Right Rear
10-369-189 No Cutouts
10-369-193 Right Side
10-369-190 Left Rear
10-369-191 Right Rear
10-369-192 Left Side
10-369-194 Left Rear, Right Rear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-369-195 Supplier Labconco™ Supplier No. 9503622
Add to Cart
Add to Cart
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
  • Dished and contoured to conform to interior liner of Protector Laboratory Hoods
  • Front edge with large radius to aerodynamically direct airflow in hood
  • Pre-drilled holes for venting and plumbing passthroughs
  • Includes left rear cupsink cutout
Specifications
For Use With (Equipment) Protector XStream Fume Hood (72 in.)
Includes Left Side, Right Rear
Product Type Work Surface
Material Epoxy Resin
For Use With (Application) Laboratory Fume Hood Work