Learn More
Labconco™ SpillStopper™ Work Surface for 8ft. Protector™Premier™and Protector™XL Hoods
GSA_VASDP
Catalog No. 10369173 Shop All Labconco Corporation Products
Change view
Click to view available options
Cut Style:
Left Rear Cupsink Cutout
Left Side Cupsink Cutout
Left and Right Rear Cupsink Cutouts
Left and Right Side Cupsink Cutouts
No Cupsink Cutout
Right Rear Cupsink Cutout
Right Side Cupsink Cutout
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cut Style
10-369-173 Left and Right Rear Cupsink Cutouts
10-369-174 Left and Right Side Cupsink Cutouts
10-369-170 Right Rear Cupsink Cutout
10-369-168 No Cupsink Cutout
10-369-171 Left Side Cupsink Cutout
10-369-172 Right Side Cupsink Cutout
10-369-169 Left Rear Cupsink Cutout
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-369-173 Supplier Labconco™ Supplier No. 9500811
Add to Cart
Add to Cart
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
SpillStopper work surfaces are dished to contain spills
  • Molded from special formulation of corrosion-resistant epoxy resins
  • Front edge has large radius to aerodynamically direct airflow into the fume hood
  • Pre-drilled holes for venting and plumbing passthroughs are provided
  • Some include 6 x 3 in. cupsink cutouts
9500820 and 9500802
Not for use with Protector Solvent Storage Cabinets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Protector XL Laboratory Hood
Product Type SpillStopper Work Surface (8 ft.)
For Use With (Application) 8 Ft. Protector Premier and Protector XL Hoods
Cut Style Left and Right Rear Cupsink Cutouts
WARNING: Cancer - www.P65Warnings.ca.gov