Learn More
Labconco™ Stainless Steel Hooks for Forensic Enclosures
SDP
Catalog No. 09681285 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Capture Portable Fuming System
Catalog No. For Use With (Equipment)
09-681-285 Capture Portable Fuming System
1 options
Catalog No. 09-681-285 Supplier Labconco™ Supplier No. 3422202
Add to Cart
Add to Cart
Accessories for use with CApture Portable Fuming System.
Allows evidence to hang inside the clear chamber during development. Type 304 stainless steel. One hook is included with each CApture Portable Fuming System.
Specifications
For Use With (Equipment) Capture Portable Fuming System
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Cyanoacrylate Fuming Chamber
Material Stainless Steel
For Use With (Application) Allows evidence to hang inside the clear chamber during development
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov