Learn More
Labconco™ Storage Shelf Kit for Precise™ Glove Boxes
SDP
Catalog No. 16108394 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box
Catalog No. For Use With (Equipment)
16-108-394 Precise Basic Glove Box
1 options
Catalog No. 16-108-394 Supplier Labconco™ Supplier No. 5235000
Add to Cart
Add to Cart
Accessories for use with Precise Glove Boxes
  • Rear interior
  • Provides additional usable surface area for small items inside glove box
  • Adds 240 sq. in. of usable space
  • Consists of 4 SS shelves, 2 epoxy-coated steel vertical uprights, SS slotted back
Specifications
For Use With (Equipment) Precise Basic Glove Box
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Includes 4 Stainless Steel Shelves, 2 Epoxy Coated Steel Vertical Uprights, Stainless Steel Slotted Back
Product Type Storage Shelf Kit
WARNING: Cancer - www.P65Warnings.ca.gov