Learn More
Labconco™ Tissue Screen Kits
SDP
Catalog No. 10304191 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Premier Hood
Protector XL Hood
Protector XStream Hood
Width (Metric):
1.21 m
1.5 m
1.8 m
2.1 m
2.4 m
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Width (Metric)
10-304-191 Protector Premier Hood 1.8 m
10-304-180 Protector XL Hood 1.21 m
10-304-181 Protector XL Hood 1.5 m
10-304-183 Protector XL Hood 2.1 m
10-304-184 Protector XL Hood 2.4 m
10-304-185 Protector XStream Hood 1.21 m
10-304-186 Protector XStream Hood 1.5 m
10-304-187 Protector XStream Hood 1.8 m
10-304-188 Protector XStream Hood 2.4 m
10-304-190 Protector Premier Hood 1.5 m
10-304-192 Protector Premier Hood 2.4 m
10-304-182 Protector XL Hood 1.8 m
10-304-189 Protector Premier Hood 1.21 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 10-304-191 Supplier Labconco™ Supplier No. 9577112
Add to Cart
Add to Cart
Mounts behind baffles for trapping tissue wipers and other papers
  • Each kit includes mounting hardware, powder-coated stainless steel screen and instructions
Specifications
For Use With (Equipment) Protector Premier Hood
Width (English) 6 ft.
Width (Metric) 1.8 m