Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Transfer Caddy
SDP

Catalog No. 10265176 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
XPert Bulk Powder Enclosures
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
10-265-176 XPert Bulk Powder Enclosures
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-265-176 Supplier Labconco™ Supplier No. 3961300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement caddy for XPert™ Bulk Powder Enclosure, Filtered System, and Station

  • Durable steel frame
  • Frame Coating: Glacier white epoxy
  • Expedites source container retrieval
  • Frame Size: 2.5cm (1 in.) square
  • Caddy latch and position plate
  • 7.6cm (3 in.) casters

Specifications

For Use With (Equipment) XPert Bulk Powder Enclosures
Weight (English) 35 lb.
Weight (Metric) 15.9 Kg
Product Type Transfer Caddy
For Use With (Application) Caddy Latch and Positioning Plate
Hood Depth (English) 30 inches
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.