Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ UV Light for Purifier™ Delta Biosafety Cabinets
SDP

Catalog No. 09039066 Shop All Labconco Corporation Products
Change view
Click to view available options
Base Style:
Single Pin Slimline (FA8)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Base Style
09-039-066 Single Pin Slimline (FA8)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-039-066 Supplier Labconco™ Supplier No. 1231900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UV lamps for Labconco Purifier Biosafety Cabinets and PuriCare Procedure Stations.

UV lamp for use with 3', 4', 5' and 6' Purifier Axiom, Logic+, Cell Logic+ and Delta Biosafety Cabinets and PuriCare Procedure Stations.

Specifications

Base Style Single Pin Slimline (FA8)
Gas Fill Argon
Light Source Germicidal UV-C
Lighting Life 7,500 hours
Voltage 100/115/220/230 V
Wattage 65 watts
Wavelength 254 nm
Weight (English) 17 lb.
Weight (Metric) 7.7 Kg
Color Clear
For Use With (Application) Secondary Disinfection
For Use With (Equipment) Purifier Delta Biosafety Cabinet
Length (English) 6 ft.
Material Borosilicate Glass
Product Type UV Bulb
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.