Learn More
Labconco™ WaterPro™Interconnect Kit
SDP
Catalog No. 16307138 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
SteamScrubber Glassware Washer
Catalog No. For Use With (Equipment)
16-307-138 SteamScrubber Glassware Washer
1 options
Catalog No. 16-307-138 Supplier Labconco™ Supplier No. 9100200
Add to Cart
Add to Cart
Connect WaterPro RO Station or 70 Liter Storage Tank to glassware washer.
The Interconnect Kit includes the necessary hardware and tubing to install a WaterPro RO Station or 70 Liter Storage Tank to a glassware washer. This kit may be used with any SteamScrubber or FlaskScrubber Glassware Washer.
Specifications
Product Type Interconnect Kit
Product Line WaterPro
For Use With (Equipment) SteamScrubber Glassware Washer
Includes Hardware and tubing to install a WaterPro™ RO station or 70L storage tank to a glassware washer
Weight (English) Shipping 5 lb.
Weight (Metric) Shipping 2.26 kg
Quantity Each