Learn More
Labconco™ WaterPro™ Softener Accessories
SDP
Catalog No. 16107161 Shop All Labconco Corporation Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
16-107-161 Tap
1 options
Catalog No. 16-107-161 Supplier Labconco™ Supplier No. 9096400
Add to Cart
Add to Cart
For use with WaterPro™ Softeners.
Replacement Color-Coded Hardness Test Strips, package of 10.
Specifications
Feedwater Source Tap
Material Mixed Cellulose Ester
For Use With (Equipment) WaterPro™ Softener
Type Replacement Hardness Test Strip
Weight (Metric) 0.2 kg
WARNING: Cancer - www.P65Warnings.ca.gov