Learn More
Labconco™ WaterPro™ Wall Mounting Panel Kit
SDP
Catalog No. 9078800 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
WaterPro PS Polishing System, WaterPro RO Systems
Catalog No. For Use With (Equipment)
90-788-00 WaterPro PS Polishing System, WaterPro RO Systems
1 options
Catalog No. 90-788-00 Supplier Labconco™ Supplier No. 9078800
Add to Cart
Add to Cart
Allows WaterPro RO and PS Systems to be hung on a wall.
Wall Mounting Panel Kit includes a stainless steel panel and hardware for mounting to the wall behind the Labconco WaterPro RO and PS Systems. Facilitates installation and helps to protect the wall from moisture.
Specifications
Type Wall Mounting Kit
For Use With (Equipment) WaterPro PS Polishing System, WaterPro RO Systems
Includes Stainless steel panel and hardware for mounting a WaterPro RO or WaterPro PS System to a flat wall surface
Width (English) 38 in.
Height (English) 37 in.
Depth (English) 10 in.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov