Learn More
Labconco™ XPert™TXE Telescoping Base Stands
Adjustable height base stands designed for Labconco XPert TXE Equipment Enclosures.
$1376.00 - $1702.00
| Catalog Number | Mfr. No. | Depth (English) | Width (English) | Weight (English) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Depth (English) | Width (English) | Weight (English) | Price | Quantity & Availability | |||
13-118-56
![]() ![]() |
Labconco™
3746715 |
29.9 in. | 24 in. | 80 lb. |
Each for $1,376.00
|
|
|||
13-118-57
![]() ![]() |
Labconco™
3746716 |
33.8 in. | 25.2 in. | 100 lb. |
Each for $1,702.00
|
|
|||
Description
XPert TXE Telescoping Base Stands support XPert TXE Equipment Enclosures. Powder-coated 1.75 in. tubular steel frame. Adjustable to eight height positions at installation in 1 in. increments from 27.5 in. to 34.5 in.. Available with fixed feet or 5 in. diameter casters.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
| 27.5 to 34.5 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.