Learn More
Description
- Label Any DNA or RNA Template – Suitable for a wide range of applications
- One-step Chemical Method – Easily and consistently control the labeling reactions
- Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Specifications
Specifications
| Detection Method | Fluorescence |
| Label or Dye | Alexa Fluor 546 |
| Final Product Type | DNA (Labeled), RNA (Labeled) |
| Sample Type | DNA/RNA |
| Labeling Target | DNA/RNA |
| Product Type | Label Reagent |
| Quantity | 100 μg |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.