Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, TM-Rhodamine

Catalog No. p-7250596 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR4100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MIR4100 Supplier Mirus Bio™ Supplier No. MIR 4100
This item is not returnable. View return policy

Label IT™ Nucleic Acid Labeling Reagents for efficient, direct, non-enzymatic labeling of DNA and RNA.

  • Label Any DNA or RNA Template – Suitable for a wide range of applications
  • One-step Chemical Method – Easily and consistently control the labeling reactions
  • Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
  • Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance

NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.

Specifications

Detection Method Fluorescence
Label or Dye Alexa Fluor 546
Final Product Type DNA (Labeled), RNA (Labeled)
Sample Type DNA/RNA
Labeling Target DNA/RNA
Product Type Label Reagent
Quantity 100 μg
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.