Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Plasmid Delivery Control, Cy3

Catalog No. MIR7904 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR7904 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MIR7904 Supplier Mirus Bio™ Supplier No. MIR 7904

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Label IT™ Plasmid Delivery Control is fluorescently labeled plasmid ideal for in vitro and in vivo tracking.

  • Sensitive – Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy
  • Compatible – Suitable for co-delivery experiments with functional plasmids
  • Ease of Use – Supplied as a ready to use 0.5 mg/mL concentration for both in vitro and in vivo tracking studies

Specifications

Concentration 0.5 mg/mL
Detection Method Fluorescence
Label or Dye Cyanine3
Final Product Type DNA
Sample Type DNA
Labeling Target DNA
Product Type Label Reagent
Quantity 10 μg
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.