Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio Label IT™ Plasmid Delivery Control, Fluorescein

Catalog No. MIR7907 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Final Product Type:
DNA (Labeled), RNA (Labeled)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Final Product Type
MIR7907 DNA (Labeled), RNA (Labeled)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MIR7907 Supplier Mirus Bio Supplier No. MIR 7907
Add to Cart
Add to Cart
This item is not returnable. View return policy

Label IT Plasmid Delivery Controls are fluorescently labeled plasmid ideal for in vitro and in vivo tracking.

  • Sensitive: Easily visualize transfected cells and assess delivery efficiency using fluorescent microscopy.
  • Compatible: Suitable for co-delivery experiments with functional plasmids.
  • Ease of Use: Supplied as a ready to use 0.5 mg/mL concentration for both in vitro and in vivo tracking studies.

Specifications

Detection Method Fluorescence
Format Kit
Label or Dye FAM
Final Product Type DNA (Labeled), RNA (Labeled)
Labeling Target DNA/RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.